3-Iodo-p-toluidine ≥97%

631264-25G 631264-5G
220082-823EA 179.85 USD
220082-823 220082-824
3-Iodo-p-toluidine ≥97%
3-Iodo-p-toluidine
Formula: C₇H₈IN
MW: 233.05 g/mol
MDL Number: MFCD00047843
CAS Number: 35944-64-0
EINECS: 252-803-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR