3-Bromo-N-methylaniline ≥97%

630985-1G 630985-5G
220082-324EA 37.69 USD
220082-324 220082-325
3-Bromo-N-methylaniline ≥97%
3-Bromo-N-methylaniline
Formula: C₇H₈BrN
MW: 186.05 g/mol
MDL Number: MFCD05664376
CAS Number: 66584-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR