4-Bromo-N-methylaniline ≥97%

630977-1G 630977-5G
220082-986EA 76.26 USD
220082-986 220082-987
4-Bromo-N-methylaniline ≥97%
4-Bromo-N-methylaniline
Formula: C₇H₈BrN
MW: 186.05 g/mol
MDL Number: MFCD03030391
CAS Number: 6911-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR