3-Fluoro-N-methylaniline ≥97%

630969-1G 630969-5G
220082-983EA 94.38 USD
220082-983 220082-984
3-Fluoro-N-methylaniline ≥97%
3-Fluoro-N-methylaniline
Formula: C₇H₈FN
MW: 125.15 g/mol
MDL Number: MFCD02683095
CAS Number: 1978-37-6
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR