2-Bromo-N-methylaniline ≥95%

630918-1G 630918-5G
220082-306EA 126 USD
220082-306 220082-307
2-Bromo-N-methylaniline ≥95%
2-Bromo-N-methylaniline
Formula: C₇H₈BrN
MW: 186.05 g/mol
MDL Number: MFCD06798064
CAS Number: 6832-87-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR