6-Aminoindole

630721-1G 630721-5G
220082-966EA 81.31 USD
220082-966 220082-967
6-Aminoindole
6-Aminoindole
Formula: C₈H₈N₂
MW: 132.16 g/mol
Boiling Pt: 161…166 °C (2.3 mmHg)
Melting Pt: 65…66 °C
Storage Temperature: Freezer
MDL Number: MFCD02093959
CAS Number: 5318-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR