Lithium chloride hydrate

Warning

62486-25G 62486-100G
220082-747EA 381.54 USD
220082-747 220082-746
Lithium chloride hydrate
Lithium chloride hydrate
Formula: ClLi·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00149764
CAS Number: 85144-11-2
EINECS: 231-212-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR