Lithium tetrahydroborate

Synonyms: Lithium tetrahydridoborate, Lithium borohydride

62460-25G-F 62460-5G-F
220082-665EA 102.46 USD
220082-665 220082-666
Lithium tetrahydroborate
Lithium tetrahydroborate
Formula: H₄BLi
MW: 21.78 g/mol
Boiling Pt: 66 °C (1013 hPa)
Melting Pt: 274…277 °C
Density: 0.666 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011088
CAS Number: 16949-15-8
EINECS: 241-021-7
UN: 1413
ADR: 4.3,I
Merck Index: 13,05547

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR