Lithium aluminum hydride

Danger

Synonyms: Aluminium lithium hydride, Lithium tetrahydridoaluminate

62420-10G-F 62420-50G-F 62420-250G-F
220081-798EA 38.08 USD
220081-798 220081-800 220081-799
Lithium aluminum hydride
Lithium aluminum hydride

Packed in plastic bag or which is soluble at 45-60 deg C in hydrocarbons or THF and dioxane

Formula: H₄AlLi
MW: 37.95 g/mol
Melting Pt: 125 °C
Density: 0.91 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011075
CAS Number: 16853-85-3
EINECS: 240-877-9
UN: 1410
ADR: 4.3,I
Merck Index: 13,00348

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR