N-Lauroylsarcosine sodium salt ≥97%

Danger

Synonyms: Lauroylsarcosine sodium, Sarkosyl, Sodium lauroyl sarcosine, Sodium N-lauroylsarcosinate, Sodium N-dodecanoyl-N-methylglycinate

61745-250G 61745-1KG 61745-50G
220081-141EA 192.77 USD
220081-141 220081-140 220081-142
N-Lauroylsarcosine sodium salt ≥97%
N-Lauroylsarcosine sodium salt
Formula: H₃C(CH₂)₁₀C(O)N(CH₃)CH₂COONa
MW: 293.38 g/mol
Melting Pt: 45 - 47 °C
Flash Pt: ≥ 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00042728
CAS Number: 137-16-6
EINECS: 205-281-5
UN: 2811
ADR: 6.1,II
Merck Index: 12,04379
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0126d80345cce5dd675a83796c379bd44cebacef&vid=48c74917d6a93a32467317ef6c6fdbf24f3b6dc2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 97%HPLC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR