Deuterium oxide

613398-10G 613398-50G
220082-351EA 60.31 USD
220082-351 220082-352
Deuterium oxide
Deuterium oxide
Formula: D₂O
Boiling Pt: 101.3 °C (1013 hPa)
Melting Pt: 3.8 °C
Density: 1.1 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00044636
CAS Number: 7789-20-0
EINECS: 232-148-9
Merck Index: 12,02984

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR