Potassium tellurite

Danger

Synonyms: Potassium tellurite, Potassium tellurium trioxide, Potassium tellurium (IV) oxide

60539-50G 60539-10G
220081-986EA 276.92 USD
220081-986 220081-985
Potassium tellurite
Potassium tellurite
Formula: K₂TeO₃
MW: 253.79 g/mol
MDL Number: MFCD00011379
CAS Number: 7790-58-1
EINECS: 232-213-1
UN: 3284
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR