3-Hydroxy-p-anisaldehyde ≥95.0%

Synonyms: Isovanillin, Isovanicaline, 3-Hydroxy-4-methoxybenzaldehyde, Isovanillic aldehyde

59940-500G 59940-100G
220081-694EA 301.54 USD
220081-694 220081-693
3-Hydroxy-p-anisaldehyde ≥95.0%
3-Hydroxy-p-anisaldehyde
Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 308 °C (1013 hPa)
Melting Pt: 68…70 °C
Density: 1.401 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003369
CAS Number: 621-59-0
EINECS: 210-694-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR