5-Bromo-2-chloropyrimidine

596949-5G 596949-1G
220081-551EA 92.4 USD
220081-551 220081-550
5-Bromo-2-chloropyrimidine
5-Bromo-2-chloropyrimidine
Formula: C₄H₂BrClN₂
MW: 193.43 g/mol
MDL Number: MFCD00483232
CAS Number: 32779-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR