2-Bromo-5-fluoropyridine

595675-5G 595675-1G
220080-835EA 100.48 USD
220080-835 220080-834
2-Bromo-5-fluoropyridine
2-Bromo-5-fluoropyridine
Formula: C₅H₃BrFN
MW: 175.99 g/mol
MDL Number: MFCD00234011
CAS Number: 41404-58-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR