ω-Bromo-2,4-dichloroacetophenone ≥97%

Synonyms: 2-Bromo-1-(2,4-dichlorophenyl)ethan-1-one, 2,4-Dichlorophenacyl bromide, α-Bromo-2,4-dichloroacetophenone, 2-Bromo-2',4'-dichloroacetophenone, 2-Bromo-1-(2,4-dichlorophenyl)ethanone

595268-1G 595268-5G
220081-424EA 50.77 USD
220081-424 220081-425
ω-Bromo-2,4-dichloroacetophenone ≥97%
ω-Bromo-2,4-dichloroacetophenone
Formula: C₈H₅BrCl₂O
MW: 267.94 g/mol
MDL Number: MFCD00053005
CAS Number: 2631-72-3
EINECS: 220-116-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR