2-Ethoxy-5-fluorophenylboronic acid

Synonyms: Dihydroxy(2-ethoxy-5-fluorophenyl)borane, (2-Ethoxy-5-fluorophenyl)boronic acid, 2-Ethoxy-5-fluorobenzeneboronic acid, (2-Ethoxy-5-fluorophenyl)metaboric acid

594954-1G 594954-10G
220081-404EA 153.46 USD
220081-404 220081-403
2-Ethoxy-5-fluorophenylboronic acid
2-Ethoxy-5-fluorophenylboronic acid
Formula: C₈H₁₀BFO₃
MW: 183.97 g/mol
MDL Number: MFCD05664328
CAS Number: 864301-27-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR