ω-Bromo-2-hydroxyacetophenone ≥97%

Synonyms: 2-Hydroxyphenacyl bromide, 2-Bromo-1-(2-hydroxyphenyl)ethan-1-one, 2-Bromo-1-(2-hydroxyphenyl)ethanone, α-Bromo-2-hydroxyacetophenone, 2-Bromo-2'-hydroxyacetophenone

594938-5G 594938-1G
220081-402EA 153.46 USD
220081-402 220081-401
ω-Bromo-2-hydroxyacetophenone ≥97%
ω-Bromo-2-hydroxyacetophenone
Formula: C₈H₇BrO₂
MW: 215.05 g/mol
MDL Number: MFCD01727570
CAS Number: 2491-36-3
UN: 3263
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR