ω-Bromo-3-hydroxyacetophenone

Synonyms: α-Bromo-3-hydroxyacetophenone, 3-Hydroxyphenacyl bromide, 2-Bromo-1-(3-hydroxyphenyl)ethan-1-one, 2-Bromo-1-(3-hydroxyphenyl)ethanone, 2-Bromo-3'-hydroxyacetophenone

594709-1G 594709-5G
220081-150EA 122.77 USD
220081-150 220081-151
ω-Bromo-3-hydroxyacetophenone
ω-Bromo-3-hydroxyacetophenone
Formula: C₈H₇BrO₂
MW: 215.05 g/mol
MDL Number: MFCD05664326
CAS Number: 2491-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR