ω-Bromo-2-chloroacetophenone ≥95%

Synonyms: α-Bromo-2-chloroacetophenone, Bromomethyl o-chlorophenyl ketone, 2-Bromo-1-(2-chlorophenyl)ethanone, 2-Bromo-2'-chloroacetophenone, 2-Chlorophenacyl bromide

594482-1G 594482-5G
220080-226EA 68.82 USD
220080-226 220080-227
ω-Bromo-2-chloroacetophenone ≥95%
ω-Bromo-2-chloroacetophenone
Formula: C₈H₆BrClO
MW: 233.49 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00832993
CAS Number: 5000-66-8
UN: 1760
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR