2-Bromo-4-fluoro-1-iodobenzene ≥97%

Warning

Synonyms: 3-Bromo-1-fluoro-4-iodobenzene, 2-Bromo-4-fluoroiodobenzene, 1-Bromo-5-fluoro-2-iodobenzene

594326-1G 594326-5G
220081-313EA 65.58 USD
220081-313 220081-314
2-Bromo-4-fluoro-1-iodobenzene ≥97%
2-Bromo-4-fluoro-1-iodobenzene
Formula: C₆H₃BrFI
MW: 300.9 g/mol
MDL Number: MFCD00142868
CAS Number: 202865-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR