5-Bromoisoquinoline ≥98%

Warning

594261-1G 594261-5G
220081-298EA 76.15 USD
220081-298 220081-299
5-Bromoisoquinoline ≥98%
5-Bromoisoquinoline
Formula: C₉H₆BrN
MW: 208.06 g/mol
MDL Number: MFCD01646405
CAS Number: 34784-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR