2-Fluoronicotinic acid

Synonyms: 2-Fluoropyridine-3-carboxylic acid, 2-Fluoro-3-pyridinecarboxylic acid

593176-1G 593176-5G
220081-217EA 200.77 USD
220081-217 220081-218
2-Fluoronicotinic acid
2-Fluoronicotinic acid
Formula: C₆H₄FNO₂
MW: 141.1 g/mol
MDL Number: MFCD00040744
CAS Number: 393-55-5
EINECS: 206-888-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR