Aluminium nitride

Synonyms: Aluminum nitride

593044-10G 593044-50G
220081-195EA 198.46 USD
220081-195 220081-196
Aluminium nitride
Aluminium nitride
Formula: AlN
MW: 40.99 g/mol
Boiling Pt: 2517 °C (1013 hPa)
Melting Pt: 1900 - 2200 °C
Density: 3.257 to 3.267 g/cm³ (160 °C)
MDL Number: MFCD00003429
CAS Number: 24304-00-5
EINECS: 246-140-8
Merck Index: 12,00366

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR