1-tert-Butyl-4-ethynylbenzene

Synonyms: 1-(tert-Butyl)-4-ethynylbenzene, (4-tert-Butylphenyl)ethyne, (4-tert-Butylphenyl)acetylene, 4-tert-Butylphenylacetylene, 1-Ethynyl-4-tert-butylbenzene

593001-25G 593001-5G
220081-186EA 288.62 USD
220081-186 220081-187
1-tert-Butyl-4-ethynylbenzene
1-tert-Butyl-4-ethynylbenzene
Formula: C₁₂H₁₄
MW: 158.24 g/mol
MDL Number: MFCD00190197
CAS Number: 772-38-3
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR