3-Phenoxybenzyl chloride ≥97%

590932-1G 590932-5G
220080-788EA 81.63 USD
220080-788 220080-789
3-Phenoxybenzyl chloride ≥97%
3-Phenoxybenzyl chloride
Formula: C₁₃H₁₁ClO
MW: 218.68 g/mol
Boiling Pt: 123 °C (0.3 -rr)
Density: 1.16 g/cm³ (20 °C)
MDL Number: MFCD00040866
CAS Number: 53874-66-1
EINECS: 258-831-1
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR