(3-Phenoxyphenyl)acetic acid ≥98%

Warning

Synonyms: 3-Phenoxyphenylacetic acid

590827-1G 590827-5G
220081-054EA 67.95 USD
220081-054 220081-055
(3-Phenoxyphenyl)acetic acid ≥98%
(3-Phenoxyphenyl)acetic acid
Formula: (C₆H₅OC₆H₅)CH₂COOH
MW: 228.25 g/mol
MDL Number: MFCD00016826
CAS Number: 32852-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR