Terbium(III) oxide ≥99.99%

590509-2G 590509-10G
220081-027EA 216.92 USD
220081-027 220081-026
Terbium(III) oxide ≥99.99%
Terbium(III) oxide
Formula: O₃Tb₂
MW: 365.85 g/mol
CAS Number: 12036-41-8
EINECS: 234-849-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR