2-Chloro-6-fluorophenol ≥98%

Danger

590169-25G 590169-5G
220081-000EA 219.69 USD
220081-000 220081-001
2-Chloro-6-fluorophenol ≥98%
2-Chloro-6-fluorophenol
Formula: C₆H₄ClFO
MW: 146.55 g/mol
Boiling Pt: 199 °C (1013 hPa)
Melting Pt: 65 °C
MDL Number: MFCD01631574
CAS Number: 2040-90-6
EINECS: 433-890-8
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR