(3-Iodophenyl)acetic acid ≥95%

Warning

Synonyms: 2-(3-Iodophenyl)acetic acid

587001-1G 587001-5G
220079-487EA 59.23 USD
220079-487 220079-488
(3-Iodophenyl)acetic acid ≥95%
(3-Iodophenyl)acetic acid
Formula: (C₆H₄I)CH₂COOH
MW: 262.05 g/mol
MDL Number: MFCD00046548
CAS Number: 1878-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR