(3-Iodopropyl)trimethoxysilane

58035-25ML-F 58035-100ML-F
220080-563EA 164.77 USD
220080-563 220080-562
(3-Iodopropyl)trimethoxysilane
(3-Iodopropyl)trimethoxysilane
Formula: IH₂CCH₂CH₂Si(OCH₃)₃
MW: 290.17 g/mol
MDL Number: MFCD00054838
CAS Number: 14867-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR