Methyl-5-chloro-2-nitrobenzoate ≥97%

Synonyms: 5-Chloro-2-nitrobenzoic acid methyl ester, Methyl 5-chloro-2-nitrobenzoate, Methyl 5-chloro-2-nitrobenzenecarboxylate

579823-5G 579823-25G
220080-541EA 33.6 USD
220080-541 220080-540
Methyl-5-chloro-2-nitrobenzoate ≥97%
Methyl-5-chloro-2-nitrobenzoate
Formula: C₈H₆ClNO₄
MW: 215.59
Boiling Pt: 306
Melting Pt: 48.5
Density: 1.426
MDL Number: MFCD00272139
CAS Number: 51282-49-6
EINECS: 257-107-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR