4-Bromo-3,5-dimethoxybenzoic acid ≥97%

579718-5G 579718-25G
220080-533EA 58.58 USD
220080-533 220080-532
4-Bromo-3,5-dimethoxybenzoic acid ≥97%
4-Bromo-3,5-dimethoxybenzoic acid
Formula: C₉H₉BrO₄
MW: 261.07 g/mol
MDL Number: MFCD01632140
CAS Number: 56518-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR