Diethylene glycol monomethyl ether ≥99.0%

Warning

Synonyms: 2-(2-Methoxyethoxy)ethanol, Diethylene glycol methyl ether

579548-1L 579548-4L
220080-521EA 21.88 USD
220080-521 220080-522
Diethylene glycol monomethyl ether ≥99.0%
Diethylene glycol monomethyl ether
Formula: C₅H₁₂O₃
MW: 120.15 g/mol
Boiling Pt: 193 °C (1013 hPa)
Melting Pt: –65 °C
Density: 1.025 g/cm³ (20 °C)
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00002871
CAS Number: 111-77-3
EINECS: 203-906-6
Merck Index: 12,06116

Order Now

Specification Test Results

Assay 99.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR