4-Iodobenzonitrile ≥97%

Synonyms: p-Iodobenzonitrile

579157-1G 579157-10G
220080-500EA 62.35 USD
220080-500 220080-499
4-Iodobenzonitrile ≥97%
4-Iodobenzonitrile
Formula: C₇H₄IN
MW: 229.02 g/mol
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 125 - 128 °C
MDL Number: MFCD00051310
CAS Number: 3058-39-7
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR