3-Cyanophenylboronic acid pinacol ester ≥97%

Synonyms: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile, 3-Cyanobenzeneboronic acid pinacol ester

578401-1G 578401-5G
220080-436EA 37.86 USD
220080-436 220080-437
3-Cyanophenylboronic acid pinacol ester ≥97%
3-Cyanophenylboronic acid pinacol ester
Formula: C₁₃H₁₆BNO₂
MW: 229.09 g/mol
MDL Number: MFCD03789262
CAS Number: 214360-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR