Hexacarbonylmolybdenum

Synonyms: Molybdenum carbonyl, Hexacarbonylmolybdenum(0), Molybdenumhexacarbonyl, Molybdenum hexacarbonyl

577766-25G 577766-5G
220079-352EA 584.62 USD
220079-352 220079-353
Hexacarbonylmolybdenum
Hexacarbonylmolybdenum
Formula: C₆MoO₆
MW: 264 g/mol
Boiling Pt: 155 °C (1013 hPa)
Density: 1.96 g/cm³ (20 °C)
MDL Number: MFCD00003466
CAS Number: 13939-06-5
EINECS: 237-713-3
UN: 3466
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR