Trifluoroacetic acid 0.1% (v/v) in aqueous solution for HPLC

576905-200L-LS 576905-4X4L 576905-18L 576905-20L
220080-384EA 1058.46 USD
220080-384 220080-386 220080-383 220080-385
Trifluoroacetic acid 0.1% (v/v) in aqueous solution for HPLC
Trifluoroacetic acid
Formula: C₂HF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD00004169
CAS Number: 76-05-1
EINECS: 200-929-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9501f3ac85d66e8d19d1cd1c3fe5409b974e9d8a&vid=be6aa627129b49ccda76006e7003b46d79b933cc&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR