2-Amino-5-bromo-4-methylpyridine

Warning

576786-25G 576786-1G 576786-5G
220080-376EA 119.54 USD
220080-376 220080-375 220080-377
2-Amino-5-bromo-4-methylpyridine
2-Amino-5-bromo-4-methylpyridine
Formula: C₆H₇BrN₂
MW: 187.04 g/mol
MDL Number: MFCD03427660
CAS Number: 98198-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR