2-Iodo-4-nitroaniline ≥97%

576689-5G 576689-1G
220080-362EA 156.92 USD
220080-362 220080-361
2-Iodo-4-nitroaniline ≥97%
2-Iodo-4-nitroaniline
Formula: C₆H₅IN₂O₂
MW: 264.02
MDL Number: MFCD00955758
CAS Number: 6293-83-0

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR