2-Aminophenylboronic acid pinacol ester

Synonyms: 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 2-Aminobenzeneboronic acid pinacol ester

576557-1G 576557-5G
220080-343EA 60.98 USD
220080-343 220080-344
2-Aminophenylboronic acid pinacol ester
2-Aminophenylboronic acid pinacol ester
Formula: C₁₂H₁₈BNO₂
MW: 219.09 g/mol
MDL Number: MFCD02179448
CAS Number: 191171-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR