3-Aminophenylboronic acid pinacol ester ≥97%

Synonyms: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 3-Aminobenzeneboronic acid pinacol ester

574686-1G 574686-5G
220080-155EA 21.88 USD
220080-155 220080-156
3-Aminophenylboronic acid pinacol ester ≥97%
3-Aminophenylboronic acid pinacol ester
Formula: C₁₂H₁₈BNO₂
MW: 219.09 g/mol
MDL Number: MFCD03453668
CAS Number: 210907-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR