3,5-Di-tert-butylbenzaldehyde ≥97%

Synonyms: 3,5-Bis(tert-butyl)benzaldehyde

574589-1G 574589-5G
220080-141EA 72.15 USD
220080-141 220080-142
3,5-Di-tert-butylbenzaldehyde ≥97%
3,5-Di-tert-butylbenzaldehyde
Formula: C₁₅H₂₂O
MW: 218.34 g/mol
MDL Number: MFCD00026298
CAS Number: 17610-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR