1-Butyl-3-methylimidazolium hydrogen sulfate, Sigma-Aldrich®

57457-1KG-F 57457-100G-F
220080-139EA 2150.06 USD
220080-139 220080-138
1-Butyl-3-methylimidazolium hydrogen sulfate, Sigma-Aldrich®
1-Butyl-3-methylimidazolium hydrogen sulphate
Formula: C₈H₁₆N₂O₄S
MW: 236.29 g/mol
MDL Number: MFCD06798197
CAS Number: 262297-13-2
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR