3-(Indol-3-yl)propionic acid

Synonyms: Indole-3-propionic acid, 3-Indolepropionic acid

57410-100G-F 57410-U 57410-25G-F
220080-119EA 105.54 USD
220080-119 220080-121 220080-120
3-(Indol-3-yl)propionic acid
3-(Indol-3-yl)propionic acid
Formula: C₁₁H₁₁NO₂
MW: 189.21 g/mol
Melting Pt: 129 - 132 °C
Density: 1.356 g/cm³ (20 °C)
MDL Number: MFCD00005660
CAS Number: 830-96-6
EINECS: 212-600-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR