Chromium(III) acetylacetonate

Danger

Synonyms: Tris(2,4-pentanedionato-O,O')chromium (III), Chromium (III) 2,4-pentanedionate, Chromium (III) tris(2,4-pentanedionato-O,O')

574082-25G 574082-5G
220080-116EA 581.54 USD
220080-116 220080-117
Chromium(III) acetylacetonate
Chromium(III) acetylacetonate
Formula: C₁₅H₂₁CrO₆
MW: 349.32 g/mol
Boiling Pt: 340 °C (1013 hPa)
Melting Pt: 215.85 °C
Density: 1.266 g/cm³ (20 °C)
MDL Number: MFCD00000015
CAS Number: 21679-31-2
EINECS: 244-526-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR