Diindium trisulfate

Synonyms: Diindium tris(sulphate), Indium(III) sulphate, Indium (III) sulphate

57151-50G 57151-10G
220079-970EA 853.85 USD
220079-970 220079-969
Diindium trisulfate
Diindium trisulphate
Formula: O₁₂In₂S₃
MW: 517.83 g/mol
Melting Pt: 585 °C
Density: 3.438 g/cm³ (20 °C)
MDL Number: MFCD00011059
CAS Number: 13464-82-9
EINECS: 236-689-1
Merck Index: 13,04978

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR