4-Fluoro-3-hydroxybenzoic acid ≥97%

571113-1G 571113-5G
220079-409EA 43.08 USD
220079-409 220079-410
4-Fluoro-3-hydroxybenzoic acid ≥97%
4-Fluoro-3-hydroxybenzoic acid
Formula: C₇H₅FO₃
MW: 156.11 g/mol
MDL Number: MFCD00269641
CAS Number: 51446-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR