Tri-tert-butylphosphine

Synonyms: Tri-t-butylphosphine, Tris(1,1-dimethylethyl)phosphine

570958-5G 570958-10G 570958-1G
220079-922EA 154.38 USD
220079-922 220079-920 220079-921
Tri-tert-butylphosphine
Tri-tert-butylphosphine
Formula: C₁₂H₂₇P
MW: 202.32 g/mol
MDL Number: MFCD00015006
CAS Number: 13716-12-6
EINECS: 237-266-4
UN: 2846
ADR: 4.2,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR