5-Bromo-2-iodopyridine

Warning

569607-5G 569607-25G
220079-767EA 69.03 USD
220079-767 220079-766
5-Bromo-2-iodopyridine
5-Bromo-2-iodopyridine
Formula: C₅H₃BrIN
MW: 283.89 g/mol
MDL Number: MFCD00691506
CAS Number: 223463-13-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR